missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ethenocytidine Monoclonal Antibody (6F5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA116886
This item is not returnable.
View return policy
Description
Specificity: This is specific for ethenocytidine and ethenodecytidine.
Ethenocytidine, also known as epsilon cytidine, is a modified nucleoside derivative of cytidine, where an etheno bridge forms between the nitrogenous base and the sugar moiety. This structural alteration is best characterized through various spectroscopic methods such as NMR, which reveal distinct differences between its crystalline and solution states. Ethenocytidine is not directly encoded by genes but can be formed post-transcriptionally as a result of exposure to certain mutagenic chemicals. The presence of ethenocytidine can induce mutagenic effects due to its propensity to mispair during DNA replication, which can alter genetic coding and affect gene expression. Understanding these modifications is crucial in the study of carcinogenesis and the design of therapeutic interventions targeting gene transcription processes.
Specifications
| Ethenocytidine | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Mouse | |
| Protein G | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG2 λ |
| ELISA, Immunocytochemistry | |
| 6F5 | |
| Unconjugated | |
| C11H13N3O5 | |
| Ethenocytidine. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| Chemical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction