Learn More
Ethopabate-d5, TRC
Specifications
Specifications
| Chemical Name or Material | Ethopabate ethyl-d5 |
| CAS | 2469244-16-2 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C12 D5 H10 N O4 |
| Quantity | 2.5 mg |
| Synonym | 4-Acetylamino-2-ethoxy-benzoic acid methyl ester-d5, 4-acetamido-2-ethoxy-benzoic acid methyl ester-d5, 4-Acetamido-2-ethoxybenzoic acid methyl ester-d5, Methyl 4-(acetylamino)-2-ethoxybenzoate-d5, Methyl 4-acetamido-2-ethoxybenzoate-d5, Methyl 2-ethoxy-4-acetamidobenzoate-d5, Ethyl Pabate-d5 |
| InChI Formula | InChI=1S/C12H15NO4/c1-4-17-11-7-9(13-8(2)14)5-6-10(11)12(15)16-3/h5-7H,4H2,1-3H3,(H,13,14)/i1D3,4D2 |
| SMILES | [2H]C([2H])([2H])C([2H])([2H])Oc1cc(NC(=O)C)ccc1C(=O)OC |
| IUPAC Name | methyl 4-acetamido-2-(1,1,2,2,2-pentadeuterioethoxy)benzoate |
| Molecular Weight (g/mol) | 242.28 |
| Show More |