missing translation for 'onlineSavingsMsg'
Learn More

(Ethoxycarbonylmethyl)triphenylphosphonium bromide, 98+%

Product Code. 11467817
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11467817 100 g 100g
11457817 25 g 25g
11477817 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11467817 Supplier Thermo Scientific Alfa Aesar Supplier No. A16347.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1530-45-6 | C22H22BrO2P | 429.29 g/mol

(Ethoxycarbonylmethyl)triphenylphosphonium bromide, is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Ethoxycarbonylmethyl)triphenylphosphonium bromide, is used as a pharmaceutical intermediate.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1530-45-6
Molecular Formula C22H22BrO2P
Molecular Weight (g/mol) 429.29
MDL Number MFCD00011835
InChI Key VJVZPTPOYCJFNI-UHFFFAOYSA-M
Synonym carbethoxymethyl triphenylphosphonium bromide, 2-ethoxy-2-oxoethyl triphenylphosphonium bromide, ethoxycarbonylmethyl triphenylphosphonium bromide, carbethoxymethylenetriphenylphosphonium bromide, ethoxycarbonylmethyl triphenyl phosphonium bromide, phosphonium, 2-ethoxy-2-oxoethyl triphenyl-, bromide, carbethoxymethyl-triphenyl-phosphonium bromide, 2-ethoxy-2-oxoethyl triphenylphosphanium bromide, carboethoxymethyl triphenylphosphonium bromide, phosphonium, ethoxycarbonylmethyl triphenyl-, bromide
PubChem CID 73731
SMILES [Br-].CCOC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∼150°C (decomposition)
Quantity 100 g
Beilstein 3581273
Sensitivity Hygroscopic
Solubility Information It is soluble in water.
IUPAC Name (2-ethoxy-2-oxoethyl)triphenylphosphanium bromide
Formula Weight 429.3
Percent Purity ≥98%
Chemical Name or Material (Ethoxycarbonylmethyl)triphenylphosphonium bromide
compliance-icons
Product Identifier
  • Carbethoxymethyl triphenylphosphonium bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Carbethoxymethyl triphenylphosphonium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.