missing translation for 'onlineSavingsMsg'
Learn More

(Ethoxycarbonylmethyl)triphenylphosphonium Bromide, TRC

Product Code. 30385854
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385854 25 mg 25mg
30381200 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30385854 Supplier Toronto Research Chemicals Supplier No. TRCE89145025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (Ethoxycarbonylmethyl)triphenylphosphonium Bromide
CAS 1530-45-6
Molecular Formula C22H22BrO2P
Quantity 25 mg
Synonym (2-Ethoxy-2-oxoethyl)(triphenyl)phosphonium Bromide, (Carbethoxymethyl)triphenylphosphonium Bromide, Carboethoxymethyltriphenylphosphonium Bromide, Ethoxycarbonylmethyltriphenylphosphonium Bromide, NSC 60450
InChI Formula InChI=1S/C22H22O2P.BrH/c1-2-24-22(23)18-25(19-12-6-3-7-13-19,20-14-8-4-9-15-20)21-16-10-5-11-17-21;/h3-17H,2,18H2,1H3;1H/q+1;/p-1
SMILES [Br-].O=C(OCC)C[P+](C=1C=CC=CC1)(C=2C=CC=CC2)C=3C=CC=CC3
Molecular Weight (g/mol) 429.29
Formula Weight 428.05
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.