Learn More
Ethyl 3,4,5-trihydroxybenzoate, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 831-61-8 |
| Molecular Formula | C9H10O5 |
| MDL Number | MFCD00016430 |
| Quantity | 100 g |
| SMILES | O=C(OCC)C1=CC(O)=C(O)C(O)=C1 |
| Molecular Weight (g/mol) | 198.17 |
| Percent Purity | 98% |
| Chemical Name or Material | Ethyl 3,4,5-trihydroxybenzoate |