missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3,4-dihydroxycinnamate, 97%

Product Code. 11476556
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11476556 1 g 1g
11486556 5 g 5g
11496556 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11476556 Supplier Thermo Scientific Alfa Aesar Supplier No. B24808.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 66648-50-8 | C11H12O4 | 208.213 g/mol

Ethyl 3,4-dihydroxycinnamate shows anti-inflammatory, immunomodulatory, and anti-carcinogenic properties. It is a potent and specific inhibitor of NF-κB and its downstream inflammatory mediators: NOS2 (inducible nitric oxide synthase; NOS; NOS II), Cox-2 (cyclooxygenase-2), and prostaglandin E2 (PGE2). Ethyl 3,4-Dihydroxycinnamate prevents DNA single-strand breaks caused by H2O2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3,4-dihydroxycinnamate shows anti-inflammatory, immunomodulatory, and anti-carcinogenic properties. It is a potent and specific inhibitor of NF-κB and its downstream inflammatory mediators: NOS2 (inducible nitric oxide synthase; NOS; NOS II), Cox-2 (cyclooxygenase-2), and prostaglandin E2 (PGE2). Ethyl 3,4-Dihydroxycinnamate prevents DNA single-strand breaks caused by H2O2.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents Store at -20°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66648-50-8
Molecular Formula C11H12O4
Molecular Weight (g/mol) 208.213
MDL Number MFCD00045754
InChI Key WDKYDMULARNCIS-GQCTYLIASA-N
Synonym ethyl caffeate, ethyl 3-3,4-dihydroxyphenyl acrylate, ethyl 3,4-dihydroxycinnamate, caffeic acid ethyl ester, ethyl trans-caffeate, unii-76gbb1ju5y, e-ethyl 3-3,4-dihydroxyphenyl acrylate, 3,4-dihydroxycinnamic acid ethyl ester, 76gbb1ju5y, chembl17347
PubChem CID 5317238
ChEBI CHEBI:69656
IUPAC Name ethyl (E)-3-(3,4-dihydroxyphenyl)prop-2-enoate
SMILES CCOC(=O)C=CC1=CC(=C(C=C1)O)O

Specifications

Quantity 1 g
Formula Weight 208.22
Percent Purity 97%
Chemical Name or Material Ethyl 3,4-dihydroxycinnamate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.