Learn More
Ethyl 3-hexenoate, 98%
Product Code. 11468617
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11468617 5 g 5g
11478617 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11468617 Supplier Thermo Scientific Alfa Aesar Supplier No. A19734.06
Request Bulk or Custom Format
CAS: 2396-83-0 | C8H14O2 | 142.20 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2396-83-0
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00036524
InChI Key VTSFIPHRNAESED-AATRIKPKSA-N
Synonym ethyl 3-hexenoate, 3-hexenoic acid, ethyl ester, ethyl hex-3-enoate, ethyl e-hex-3-enoate, 3-hexenoic acid, ethyl ester, e, ethyl trans-3-hexenoate, ethyl e-3-hexenoate, e-3-hexenoic acid ethyl ester, ethyl hydrosorbate, 3-hexenoic acid, ethyl ester, 3e
PubChem CID 5362622
IUPAC Name ethyl (E)-hex-3-enoate
SMILES CCOC(=O)C\C=C\CC
Specifications
Density 0.89
Boiling Point 63°C to 64°C (12 mmHg)
Flash Point 59°C (139°F)
Refractive Index 1.426
Quantity 5 g
UN Number UN3272
Formula Weight 142.2
Percent Purity 98%
Chemical Name or Material Ethyl 3-hexenoate