Learn More
Ethyl 3-(Methyl-d3-mercapto)propionate, TRC
Specifications
Specifications
| Chemical Name or Material | Ethyl 3-(Methyl-d3-mercapto)propionate |
| CAS | 1020719-41-8 |
| Molecular Formula | C6 2H3 H9 O2 S |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C6H12O2S/c1-3-8-6(7)4-5-9-2/h3-5H2,1-2H3/i2D3 |
| SMILES | [2H]C([2H])([2H])SCCC(=O)OCC |
| IUPAC Name | ethyl 3-(trideuteriomethylsulfanyl)propanoate |
| Molecular Weight (g/mol) | 151.24 |
| Formula Weight | 151.0746 |