Learn More
Ethyl 3-Oxododecanoate, TRC
Specifications
Specifications
| Chemical Name or Material | Ethyl 3-Oxododecanoate |
| CAS | 67342-99-8 |
| Molecular Formula | C14 H26 O3 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C14H26O3/c1-3-5-6-7-8-9-10-11-13(15)12-14(16)17-4-2/h3-12H2,1-2H3 |
| SMILES | CCCCCCCCCC(=O)CC(=O)OCC |
| IUPAC Name | ethyl 3-oxododecanoate |
| Molecular Weight (g/mol) | 242.354 |
| Formula Weight | 242.188 |