missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 4-Aminobenzoate-2,3,5,6-d4, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Benzocaine-d4 |
| CAS | 342611-08-9 |
| Molecular Formula | C9 2H4 H7 N O2 |
| Quantity | 0.05 g |
| Synonym | Benzoic-2,3,5,6-d4 acid, 4-amino-, ethyl ester (9 CI), Benzocaine-d4, Benzocaine-d4 (phenyl D4) |
| InChI Formula | InChI=1 S/C9H11NO2/c1-2-12-9(11)7-3-5-8(10)6-4-7/h3-6 H,2,10H2,1H3/i3D,4 D,5 D,6 D |
| SMILES | [2 H]c1c([2 H])c(C(=O)OCC)c([2 H])c([2 H])c1N |
| IUPAC Name | ethyl 4-amino-2,3,5,6-tetradeuteriobenzoate |
| Molecular Weight (g/mol) | 169.21 |
| Formula Weight | 169.1041 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction