Learn More
ethyl (4-nitrophenyl)carbamate, TRC
Specifications
Specifications
| Chemical Name or Material | ethyl (4-nitrophenyl)carbamate |
| CAS | 2621-73-0 |
| Molecular Formula | C9H10N2O4 |
| Quantity | 2.5 g |
| InChI Formula | InChI=1S/C9H10N2O4/c1-2-15-9(12)10-7-3-5-8(6-4-7)11(13)14/h3-6H,2H2,1H3,(H,10,12) |
| SMILES | CCOC(=O)Nc1ccc(cc1)[N+](=O)[O-] |
| Molecular Weight (g/mol) | 210.18 |
| Formula Weight | 210.0641 |