Learn More
Ethyl 8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate, 95%, Thermo Scientific™
Product Code. 15438985
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15438985 1 g 1g
15448985 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15438985 Supplier Thermo Scientific Alfa Aesar Supplier No. H35098.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1363404-84-5
Molecular Formula C11H9F3N2O2
Molecular Weight (g/mol) 258.2
MDL Number MFCD16987556
InChI Key WZLPRVSGJRYTIQ-UHFFFAOYSA-N
Synonym ethyl 8-trifluoromethyl imidazo 1,2-a pyridine-2-carboxylate, ethyl8-trifluoromethyl imidazo 1,2-a pyridine-2-carboxylate, 8-trifluoromethyl imidazo 1,2-a pyridine-2-carboxylic acid ethyl ester, 8-trifluoromethyl-imidazo 1,2-a pyridine-2-carboxylic acid ethyl ester
PubChem CID 71627091
IUPAC Name ethyl 8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
SMILES CCOC(=O)C1=CN2C=CC=C(C2=N1)C(F)(F)F
Specifications
Melting Point 96°C to 99°C
Quantity 1 g
Formula Weight 258.2
Percent Purity 95%
Chemical Name or Material Ethyl 8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate