missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl-d5-malonic Acid, CDN
missing translation for 'orderingAttributeHoverText'
Quantity:
0.01 g
0.05 g
missing translation for 'unitSize'
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Ethylmalonic Acid-D5 (ethyl-D5) |
| CAS | 66311-22-6 |
| Molecular Formula | C5 D5 H3 O4 |
| Quantity | 0.01 g |
| Synonym | Propanedioic acid, ethyl-d5- (9 CI), Ethyl-d5-malonic Acid, Ethyl-d5-propanedioic acid, 2-(Ethyl-d5)propanedioic acid, 1,1-Propanedicarboxylic-2,2,3,3,3-d5 acid, 2-(Ethyl-1,1,2,2,2-d5)malonic acid, Ethylmalonic Acid-D5 (ethyl-D5) |
| InChI Formula | InChI=1 S/C5H8O4/c1-2-3(4(6)7)5(8)9/h3H,2H2,1H3,(H,6,7)(H,8,9)/i1D3,2D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C(C(=O)O)C(=O)O |
| IUPAC Name | 2-(1,1,2,2,2-pentadeuterioethyl)propanedioic acid |
| Molecular Weight (g/mol) | 137.1454 |
| Formula Weight | 137.0736 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction