missing translation for 'onlineSavingsMsg'
Learn More

Ethyl phenyl sulfide, 98%

Product Code. p-7021588
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11491748 5 g 5g
11401758 25 g 25g
11411758 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11491748 Supplier Thermo Scientific Alfa Aesar Supplier No. A13987.06
Add to basket
This item is not returnable. View return policy

CAS: 622-38-8 | C8H10S | 138.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-38-8
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.23
MDL Number MFCD00009265
InChI Key AEHWKBXBXYNPCX-UHFFFAOYSA-N
Synonym ethyl phenyl sulfide, ethylthio benzene, benzene, ethylthio, thiophenetole, phenyl ethyl sulfide, ethylphenylsulfide, phenylthio ethane, ethylsulfanyl benzene, sulfide, ethyl phenyl, 1-thiapropyl benzene
PubChem CID 12144
IUPAC Name ethylsulfanylbenzene
SMILES CCSC1=CC=CC=C1

Specifications

Density 1.02
Boiling Point 204°C to 205°C
Flash Point 73°C (163°F)
Odor Stench
Refractive Index 1.5655
Quantity 5 g
Beilstein 1905731
Formula Weight 138.23
Percent Purity 98%
Chemical Name or Material Ethyl phenyl sulfide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.