missing translation for 'onlineSavingsMsg'
Learn More

Ethyl trans-2-decenoate, 97%, Thermo Scientific Chemicals

Product Code. 11412998
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11412998 10 g 10g
11422998 50 g 50g
11478920 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11412998 Supplier Thermo Scientific Alfa Aesar Supplier No. A18718.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7367-88-6
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.306
MDL Number MFCD00015299
InChI Key GNJARWZWODMTDR-ZHACJKMWSA-N
Synonym ethyl trans-2-decenoate, ethyl 2-decenoate, ethyl e-2-decenoate, trans-2-decenoic acid ethyl ester, ethyl 2e-dec-2-enoate, 2-decenoic acid, ethyl ester, 2e, unii-b2k05s6l5q, 2-decenoic acid, ethyl ester, e, ethyl 2-decenoate, 2e, fema no. 3641
PubChem CID 5463904
ChEBI CHEBI:87324
IUPAC Name ethyl (E)-dec-2-enoate
SMILES CCCCCCCC=CC(=O)OCC

Specifications

Density 0.88
Boiling Point 251°C
Flash Point 110°C (230°F)
Refractive Index 1.446
Quantity 10 g
Beilstein 1704867
Formula Weight 198.31
Percent Purity 97%
Chemical Name or Material Ethyl trans-2-decenoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.