Learn More
Ethyl trans-2-decenoate, 97%, Thermo Scientific Chemicals
Product Code. 11478920
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478920 100 g 100g
11412998 10 g 10g
11422998 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11478920 Supplier Thermo Scientific Alfa Aesar Supplier No. A18718.22
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7367-88-6
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.306
MDL Number MFCD00015299
InChI Key GNJARWZWODMTDR-ZHACJKMWSA-N
Synonym ethyl trans-2-decenoate, ethyl 2-decenoate, ethyl e-2-decenoate, trans-2-decenoic acid ethyl ester, ethyl 2e-dec-2-enoate, 2-decenoic acid, ethyl ester, 2e, unii-b2k05s6l5q, 2-decenoic acid, ethyl ester, e, ethyl 2-decenoate, 2e, fema no. 3641
PubChem CID 5463904
ChEBI CHEBI:87324
IUPAC Name ethyl (E)-dec-2-enoate
SMILES CCCCCCCC=CC(=O)OCC
Specifications
Density 0.88
Boiling Point 251°C
Flash Point 110°C (230°F)
Refractive Index 1.446
Quantity 100 g
Beilstein 1704867
Formula Weight 198.31
Percent Purity 97%
Chemical Name or Material Ethyl trans-2-decenoate