missing translation for 'onlineSavingsMsg'
Learn More

Ethyl trans-3-(1-pyrrolidinyl)acrylate, 98+%, Thermo Scientific™

Product Code. 11369125
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11369125 5 g 5g
11359125 1 g 1g
11379125 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11369125 Supplier Thermo Scientific Alfa Aesar Supplier No. L16361.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65651-80-1
Molecular Formula C9H15NO2
Molecular Weight (g/mol) 169.22
MDL Number MFCD01075735
InChI Key VFFHLZJSZHWXAJ-UHFFFAOYSA-N
Synonym ethyl trans-3-1-pyrrolidinyl acrylate, e-ethyl 3-pyrrolidin-1-yl acrylate, ethyl 2e-3-pyrrolidin-1-yl prop-2-enoate, 3-pyrrolidin-1-yl-acrylic acid ethyl ester, hcnd@nadfu^u zfjjaap, 2-propenoic acid, 3-1-pyrrolidinyl-, ethyl ester, 2e, 3-pyrrolizinoacrylic acid ethyl ester, ethyl 2e-3-pyrrolidin-1-ylacrylate, ethyl 2e-3-pyrrolidinylprop-2-enoate, ethyl e-3-pyrrolidin-1-ylprop-2-enoate
PubChem CID 639785
SMILES CCOC(=O)C=CN1CCCC1

Specifications

Melting Point 39°C to 43°C
Boiling Point 118°C (1 mmHg)
Quantity 5 g
Beilstein 4399757
IUPAC Name ethyl (E)-3-pyrrolidin-1-ylprop-2-enoate
Formula Weight 169.22
Percent Purity ≥98%
Chemical Name or Material Ethyl trans-3-(1-pyrrolidinyl)acrylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.