Learn More
Ethyl trans-4-decenoate, 97%
Product Code. 11473967
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11473967 10 g 10g
11483967 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11473967 Supplier Thermo Scientific Alfa Aesar Supplier No. A19364.09
Request Bulk or Custom Format
CAS: 76649-16-6 | C12H22O2 | 198.31 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 76649-16-6
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.31
MDL Number MFCD00015574
InChI Key AWNIQMQADACLCJ-CMDGGOBGSA-N
Synonym ethyl trans-4-decenoate, ethyl e-4-decenoate, ethyl 4-decenoate, 4-decenoic acid, ethyl ester, e, ethyl 4e-decenoate, fema no. 3642, ethyl 4e-dec-4-enoate, ethyl trans-dec-4-enoate, ethyl e-dec-4-enoate, ethyl 4e-4-decenoate
PubChem CID 5362583
IUPAC Name ethyl (E)-dec-4-enoate
SMILES CCCCC\C=C\CCC(=O)OCC
Specifications
Refractive Index 1.437
Quantity 10 g
Formula Weight 198.31
Percent Purity 97%
Chemical Name or Material Ethyl trans-4-decenoate