Learn More
Ethylbenzene-d10, CDN
Specifications
Specifications
| Chemical Name or Material | Ethylbenzene-d10 |
| CAS | 25837-05-2 |
| Molecular Formula | C8 2H10 |
| Quantity | 2.5 g |
| Synonym | Benzene-1,2,3,4,5-d5, 6-(ethyl-1,1,2,2,2-d5)-, Benzene-d5, ethyl-d5- (8 CI,9 CI), 6-(Ethyl-1,1,2,2,2-d5)benzene-1,2,3,4,5-d5, (Ethyl-d5)benzene-d5, Ethylbenzene-d10, Perdeuteroethylbenzene |
| InChI Formula | InChI=1 S/C8H10/c1-2-8-6-4-3-5-7-8/h3-7 H,2H2,1H3/i1D3,2D2,3 D,4 D,5 D,6 D,7 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,2,3,4,5-pentadeuterio-6-(1,1,2,2,2-pentadeuterioethyl)benzene |
| Molecular Weight (g/mol) | 116.23 |
| Formula Weight | 116.141 g/mol |
| Show More |