missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ethyle2-amino-4-methylpyrimidine-5-carboxylate, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 181.19 |
Percent Purity | 97% |
Chemical Name or Material | Ethyl 2-amino-4-methylpyrimidine-5-carboxylate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'