Learn More
Ethylene-d4-diamine-1-N-t-BOC, CDN
Click to view available options
Quantity:
0.1 g
Unit Size:
0.10g
Specifications
Specifications
| Chemical Name or Material | mono-BOC-ethylenediamine-1,1,2,2-D4 |
| CAS | 509148-73-6 |
| Molecular Formula | C7 D4 H12 N2 O2 |
| Quantity | 0.1 g |
| Synonym | Carbamic acid, (2-aminoethyl-1,1,2,2-d4)-, 1,1-dimethylethyl ester (9 CI), (2-Amino-ethyl-1,1,2,2-d4)-carbamic acid tert-butyl ester, N-(tert-Butyloxycarbonyl)-1,2-ethylenediamine-1,1,2,2-d4, N-tert-Butoxycarbonylethylenediamine-d4, mono-BOC-ethylenediamine-1,1,2,2-D4, Ethylene-d4-diamine-1-N-t-BOC |
| InChI Formula | InChI=1 S/C7H16N2O2/c1-7(2,3)11-6(10)9-5-4-8/h4-5,8H2,1-3H3,(H,9,10)/i4D2,5D2 |
| SMILES | [2 H]C([2 H])(N)C([2 H])([2 H])NC(=O)OC(C)(C)C |
| IUPAC Name | tert-butyl N-(2-amino-1,1,2,2-tetradeuterioethyl)carbamate |
| Molecular Weight (g/mol) | 164.2388 |
| Formula Weight | 164.1463 g/mol |
| Show More |