missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic acid tripotassium salt dihydrate, 99%

Product Code. p-7045788
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11348545

Brand: Thermo Scientific Alfa Aesar L11991.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 65501-24-8 | C10H17K3N2O10 | 442.545 g/mol

Ethylenediaminetetraacetic acid tripotassium salt dihydrate is involved to eliminate inhibition of enzyme catalyzed reactions due to traces of heavy metals. It is also used to study chelators, biochemicals and reagents. Further, it is utilized to check the safety of ethylenediaminetetraacetic acid and hydroxyethyenediaminetriacetic acid ingredients in cosmetic formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylenediaminetetraacetic acid tripotassium salt dihydrate is involved to eliminate inhibition of enzyme catalyzed reactions due to traces of heavy metals. It is also used to study chelators, biochemicals and reagents. Further, it is utilized to check the safety of ethylenediaminetetraacetic acid and hydroxyethyenediaminetriacetic acid ingredients in cosmetic formulations.

Solubility
Soluble in water and potassium hydroxide.

Notes
Incompatible with strong oxidizing agents, metals, zinc, aluminum, carbon steel, copper, copper alloys, galvanized metals and nickel.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65501-24-8
Molecular Formula C10H17K3N2O10
Molecular Weight (g/mol) 442.545
MDL Number MFCD00149672
InChI Key MAPFUJCWRWFQIY-UHFFFAOYSA-K
Synonym ethylenediaminetetraacetic acid tripotassium salt dihydrate, edta tripotassium salt, edta tripotassium salt dihydrate, tripotassium ethylenediaminetetraacetate dihydrate, potassium 2,2'-2-carboxylatomethyl carboxymethyl amino ethyl azanediyl diacetate dihydrate, tripotassium 2-2-bis carboxylatomethyl amino ethyl carboxymethyl amino acetate dihydrate, edta tripotassium salt, dihydrate, ethylenediaminetetraacetic acid, tripotassium salt dihydrate, ethylenediaminetetraacetic acid tripotassium salt dihydrate titration
PubChem CID 2733162
IUPAC Name tripotassium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxymethyl)amino]acetate;dihydrate
SMILES C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)[O-].O.O.[K+].[K+].[K+]

Specifications

Melting Point ∽182°C (decomposition)
Quantity 50 g
Beilstein 6762497
Solubility Information Soluble in water and potassium hydroxide.
Formula Weight 442.55 (406.46 Anhydrous)
Percent Purity 99%
Chemical Name or Material Ethylenediaminetetraacetic acid tripotassium salt dihydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.