Learn More
Ethylmethyl-d3-amine, CDN
Specifications
Specifications
| Chemical Name or Material | N-Ethyl-N-methylamine-D3 (N-methyl-D3) |
| CAS | 223459-64-1 |
| Molecular Formula | C3 D3 H6 N |
| Quantity | 1 g |
| Synonym | Ethanamine, N-(methyl-d3)- (9 CI), Ethylmethyl-d3-amine, N-(Methyl-d3)ethanamine, N-Ethyl-N-(methyl-d3)amine, N-Ethyl-N-methylamine-D3 (N-methyl-D3) |
| InChI Formula | InChI=1 S/C3H9N/c1-3-4-2/h4H,3H2,1-2H3/i2D3 |
| SMILES | [2 H]C([2 H])([2 H])NCC |
| IUPAC Name | N-(trideuteriomethyl)ethanamine |
| Molecular Weight (g/mol) | 62.1288 |
| Formula Weight | 62.0923 g/mol |
| Show More |