missing translation for 'onlineSavingsMsg'
Learn More

Ethynodiol diacetate, MedChemExpress

Product Code. 16422256
Encompass_Preferred
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
This item is not returnable. View return policy

Product Code. 16422256

Brand: Medchem Express HYB1089

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ethynodiol diacetate (Ethynodiol acetate) is a steroidal progestin which is used as a hormonal contraceptive, it has relatively little or no potency as an androgen,has significant estrogenic effects.

Ethynodiol diacetate (Ethynodiol acetate) is a steroidal progestin which is used as a hormonal contraceptive, it has relatively little or no potency as an androgen,has significant estrogenic effects.

Specifications

Chemical Name or Material Ethynodiol diacetate
CAS 297-76-7
Color White
Purity Grade Notes Research
Molecular Formula C24H32O4
Quantity 500 mg
Synonym Ethynodiol acetate
Solubility Information DMSO : 150 mg/mL (390.11 mM; Need ultrasonic)
SMILES C#C[C@]1(OC(C)=O)CC[C@@]2([H])[C@]3([H])CCC4=C[C@@H](OC(C)=O)CC[C@]4([H])[C@@]3([H])CC[C@]12C
Molecular Weight (g/mol) 384.51
Formula Weight 384.51
Percent Purity 98.58%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.