Learn More
Ethynyl Estradiol, MedChemExpress
Product Code. 16458176
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Product Code. 16458176
Brand: Medchem Express HYB0216
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress Ethynyl Estradiol (17α-Ethynylestradiol;Ethynylestradiol) is an orally bio-active estrogen used in almost all modern formulations of combined oral contraceptive pills.
Ethynyl Estradiol (17α-Ethynylestradiol
Specifications
Chemical Name or Material Ethynyl Estradiol
CAS 57-63-6
Color White
Purity Grade Notes Research
Molecular Formula C20H24O2
Quantity 100 mg
Synonym 17α-Ethynylestradiol Ethynylestradiol
Solubility Information DMSO : 250 mg/mL (843.45 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES OC1=CC=C2C(CC[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@]4(C#C)O)=C1
Molecular Weight (g/mol) 296.4
Formula Weight 296.4
Percent Purity 99.28%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only