Learn More
Etifoxine, MedChemExpress
Product Code. 16469627
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16469627
Brand: Medchem Express HY16579A
MedChemExpress Etifoxine, a non-benzodiazepine GABAergic compound, is a positive allosteric modulator of α1β2γ2 and α1β3γ2 subunit-containing GABAA receptors. Etifoxine reveals anxiolytic and anticonvulsant properties in rodents.
Etifoxine, a non-benzodiazepine GABAergic compound, is a positive allosteric modulator of α1β2γ2 and α1β3γ2 subunit-containing GABAA receptors. Etifoxine reveals anxiolytic and anticonvulsant properties in rodents.
Specifications
Chemical Name or Material Etifoxine
Physical Form Solid
CAS 21715-46-8
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C17H17ClN2O
For Use With (Application) Neuroscience-Neuromodulation
Synonym HOE 36-801
Solubility Information DMSO : 100 mg/mL (332.47 mM; Need ultrasonic)
SMILES ClC1=CC=C2C(C(C3=CC=CC=C3)(C)OC(NCC)=N2)=C1
Molecular Weight (g/mol) 300.78
Formula Weight 300.78
Percent Purity 99.87%
Grade Research
Show More Show Less
Research purposes only