Learn More
Invitrogen™ Etigilimab Recombinant Monoclonal Antibody
GREENER_CHOICE
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA559815
 View more versions of this product
Product Code. 30282891
  • € 500.00 / 100µg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Etigilimab is a human-derived monoclonal antibody. Etigilimab targets TIGIT. It started clinical research for the treatment of advanced solid tumors in the United States in 2017 and is currently in clinical phase I.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Etigilimab
Recombinant Monoclonal
Unconjugated
OMP-313M32
Protein A
RUO
Human
Antibody
IgG1
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized