missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ETK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104362
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total BMX.
Bone marrow tyrosine kinase (BMX) is a non-receptor tyrosine kinase that belongs to the SRC-related TEC subfamily of tyrosine kinases. It is an important regulator of various cellular processes including apoptosis, cell survival, cellular differentiation, cell migration, and transformation. One family of nonreceptor TKs includes the genes TEC (MIM 600583), TXK (MIM 600058), ITK (MIM 186973), and BTK (MIM 300300). All of these proteins are homologs of the Drosophila Src28 TK and contain an SH3 and SH2 domain upstream of the TK domain.[supplied by OMIM].
Specifications
| ETK | |
| Polyclonal | |
| Unconjugated | |
| BMX | |
| BMX; BMX non-receptor tyrosine kinase; bone marrow tyrosine kinase gene in chromosome X protein; Bone marrow tyrosine kinase gene in chromosome X protein homolog; BTK-like on X chromosome; cytoplasmic tyrosine-protein kinase BMX; epa3; Epithelial and endothelial tyrosine kinase; ETK; Etk/Bmx; Etk/Bmx cytosolic tyrosine kinase; NTK38; NTK38 tyrosine kinase; PSCTK2; PSCTK3; RGD1565643; Tyro8; TYRO8 protein tyrosine kinase 8 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 12169, 367786, 660 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P51813, P97504 | |
| BMX | |
| A synthesized peptide derived from human BMX(Accession P51813), corresponding to amino acid residues C542-K592. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction