Learn More
Etofibrate, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | Etofibrate |
| CAS | 31637-97-5 |
| Molecular Formula | C18H18ClNO5 |
| Quantity | 50 mg |
| Synonym | Nicotinic acid, 2-hydroxyethyl ester 2-(p-chlorophenoxy)-2-methylpropionate (ester) (8CI), Ethofibrate, Etofibrate, Lipo-Merz, Tricerol |
| InChI Formula | InChI=1S/C18H18ClNO5/c1-18(2,25-15-7-5-14(19)6-8-15)17(22)24-11-10-23-16(21)13-4-3-9-20-12-13/h3-9,12H,10-11H2,1-2H3 |
| SMILES | CC(C)(Oc1ccc(Cl)cc1)C(=O)OCCOC(=O)c2cccnc2 |
| IUPAC Name | 2-[2-(4-chlorophenoxy)-2-methylpropanoyl]oxyethyl pyridine-3-carboxylate |
| Molecular Weight (g/mol) | 363.79 |
| Formula Weight | 363.0874 |