missing translation for 'onlineSavingsMsg'
Learn More

Ets-1 Antibody (ETS1/1801), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-54514B

Product Code. 18635208

  • € 560.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Ets-1 Monoclonal antibody specifically detects Ets-1 in Human, Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array, CyTOF
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ets-1
Monoclonal
Biotin
Avian erythroblastosis virus E26 (v-ets) oncogene homolog-1, ets protein, ETS-1, EWSR2, FLJ10768, p54, protein C-ets-1, v-ets avian erythroblastosis virus E2 oncogene homolog 1, v-ets avian erythroblastosis virus E26 oncogene homolog 1, v-ets erythroblastosis virus E26 oncogene homolog 1 (avian)
Recombinant human Ets-1 fragment (around aa137-230) (exact sequence is proprietary) (Uniprot: P14921)
100 μg
Cancer, Cell Cycle and Replication, Chromatin Research, Innate Immunity, Transcription Factors and Regulators
2113
Store at 4°C in the dark.
IgG2b κ
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array, CyTOF
ETS1/1801
PBS
Mouse
Protein A or G purified
RUO
Primary
Human, Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.