missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETS1 associated protein II Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-247MFV610
This item is not returnable.
View return policy
Description
ETS1 associated protein II Polyclonal antibody specifically detects ETS1 associated protein II in Human samples. It is validated for Western Blot
Specifications
| ETS1 associated protein II | |
| Polyclonal | |
| Western Blot | |
| AD022, dJ30M3.3, EAP2, EAPII, EC 3.1.4.-, ETS1-associated protein 2, ETS1-associated protein II, MGC111021,5'-Tyr-DNA phosphodiesterase, MGC9099,5'-tyrosyl-DNA phosphodiesterase, TRAF and TNF receptor associated protein, TTRAPTRAF and TNF receptor-associated protein, Tyr-DNA phosphodiesterase 2, tyrosyl-DNA phosphodiesterase 2 | |
| A synthetic peptide made to residues 11-28 (REAAEEEGEPEVKKRRLL) of human EAPII. | |
| 0.1 mL | |
| Signal Transduction, Transcription Factors and Regulators | |
| 51567 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction