Learn More
ETS2 Mouse anti-Human, Clone: OTI3C4, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF505621
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ETS transcriptions factors, such as ETS2, regulate numerous genes and are involved in stem cell development, cell senescence and death, and tumorigenesis. The conserved ETS domain within these proteins is a winged helix-turn-helix DNA-binding domain that recognizes the core consensus DNA sequence GGAA/T of target genes.Specifications
| ETS2 | |
| Monoclonal | |
| Unconjugated | |
| ETS2 | |
| ETS2IT1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2114 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI3C4 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P15036 | |
| ETS2 | |
| Human recombit protein fragment corresponding to amino acids 90-349 of human ETS2 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.