missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ETS2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528053
This item is not returnable.
View return policy
Description
Recommended positive controls: HeLa. Predicted reactivity: Mouse (87%), Rat (88%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ETS transcriptions factors, such as ETS2, regulate numerous genes and are involved in stem cell development, cell senescence and death, and tumorigenesis. The conserved ETS domain within these proteins is a winged helix-turn-helix DNA-binding domain that recognizes the core consensus DNA sequence GGAA/T of target genes.
Specifications
| ETS2 | |
| Polyclonal | |
| Unconjugated | |
| ETS2 | |
| AU022856; avian erythroblastosis virus E26 (v-ets) oncogene homolog 2; E26 avian leukemia oncogene 2, 3' domain; ETS proto-oncogene 2, transcription factor; Ets2; Ets-2; ETS2IT1; oncogene ETS-2; protein C-ets-2; v-ets avian erythroblastosis virus E2 oncogene homolog 2; v-ets avian erythroblastosis virus E26 oncogene homolog 2; v-ets erythroblastosis virus E26 oncogene homolog 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2114, 23872, 304063 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.26 mg/mL | |
| PBS with 20% glycerol, 1% BSA and 0.025% ProClin 300; pH 7 | |
| P15036, P15037 | |
| ETS2 | |
| Recombinant protein encompassing a sequence within the center region of human ETS2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction