Learn More
ETV1 Antibody (CL15257) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-44305
Product Code. 30293598
  • € 638.00 / 100µL
Description
ETV1 Monoclonal antibody specifically detects ETV1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ETV1
Monoclonal
LYOPH
Western Blot 1 μg/mL
DKFZp781L0674, ER81ets variant gene 1, ETS translocation variant 1, ets variant 1, Ets-related protein 81, MGC104699, MGC120533, MGC120534
This antibody was generated using a synthetic peptide of P50549, with the exact immunogen sequence remaining proprietary.
100 μL
Stem Cell Markers
2115
Human
Purified
Western Blot
CL15257
Unconjugated
Lyophilized in 5% Trehalose.
Mouse
Protein A purified
RUO
Primary
Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL.
Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles.
IgG2b