missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ETV1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA577975
This item is not returnable.
View return policy
Description
Positive Control: A431, Rat2 Predicted Reactivity: Xenopus laevis (86%), Chicken (92%), Chimpanzee (100%), Bovine (95%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ETV1: ets variant gene 1. Several members of the Ets gene family encode sequence-specific DNA binding proteins that recognize DNA sequences with a centrally located 5'-GGAA-3' element. All of the Ets proteins recognize the same central core sequence but each protein interacts with unique sequences that flank this core. ER81 (also designated ETV1) binds the motif 5'-CGGAA/T-3'. ER81 is highly expressed in brain, testis, lung and heart. ER81 is also moderately expressed in spleen, pancreas, colon and small intestine. During development, ER81 displays a unique expression patterns which suggests that this transcriptional factor might play an important role in organogenesis. ERK-1 activates ER81 transcriptional activity while MAPKAP kinase 2 inhibits ER81.
Specifications
| ETV1 | |
| Polyclonal | |
| Unconjugated | |
| Etv1 | |
| DKFZp781L0674; Er81; ets related protein 81; ETS transcription factor Er81; ETS translocation variant 1; ets variant 1; ets variant gene 1; ETS variant transcription factor 1; ets-related protein 81; Etsrp81; Etv1; MGC104699; MGC120533; MGC120534; MGC137631 protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14009, 2115, 362733 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.34 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P41164, P50549 | |
| Etv1 | |
| Recombinant protein encompassing a sequence within the center region of human ER81. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction