missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ETV2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568763
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Dog: 77%; Human: 85%; Mouse: 100%; Rat: 93%.
Binds to DNA sequences containing the consensus pentanucleotide 5'-CGGA[AT]-3'.
Specifications
| ETV2 | |
| Polyclonal | |
| Unconjugated | |
| ETV2 | |
| ER71; ets related protein 71; ETS translocation variant 2; ETS variant 2; ets variant gene 2; ETS variant transcription factor 2; ets-related protein 71; Etsrp71; ETV2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14008 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P41163 | |
| ETV2 | |
| synthetic peptide corresponding to a region of mouse ETV2 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction