Learn More
Novus Biologicals™ ETV3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals™ NBP3-28967
Product Code. 30047931
  • € 605.00 / 100µg
Description
ETV3 Polyclonal antibody specifically detects ETV3 in Human samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ETV3
Polyclonal
Tris-citrate/phosphate buffer, pH 7 to 8
Rabbit
Affinity purified
RUO
2117
Store at 4°C. Do not freeze.
IgG
Immunoprecipitation, Western Blot
Unconjugated
bA110J1.4, ETS domain transcriptional repressor PE1, ETS translocation variant 3, ets variant 3, ets variant gene 3, ETS family transcriptional repressor, METS, Mitogenic Ets transcriptional suppressor, PE-1ets variant gene 3, PE1FLJ79173
Between 450 and 500