missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ETV4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA513595
This item is not returnable.
View return policy
Description
ETV4 Polyclonal Antibody for Western Blot, IHC (P), Flow
ETV4: ets variant gene 4 (E1A enhancer binding protein, E1AF), also known as PEA3. Several members of the Ets gene family are known to encode sequencespecific DNA binding proteins. These include mouse PU. 1, mouse and human Ets-1, Drosophila E74, chicken and human Ets-2 and rat GABP-a. Each of these proteins recognizes similar motifs in DNA that share a centrally located 5'-GGAA-3' element. For instance, PEA3 binds the motif 5'-AGGAAG-3' (the PEA-3 motif), but does not bind to the sequence 5'-AGGAAC-3', recognized by PU. 1, although PU. 1 binds equally well to both sequences. It appears that all of the Ets proteins recognize the same central core sequence but that each protein interacts with unique sequences that flank this core. PEA3 is expressed at readily detectable levels in cells of epithelial and fibroblastic origin but is not expressed in hematopoietic cells. This is in contrast to other members of the Ets gene family, such as Ets-1, Ets-2 and Fli-1, each of which is expressed primarily in cells of hematopoietic origin.
Specifications
| ETV4 | |
| Polyclonal | |
| Unconjugated | |
| ETV4 | |
| Adenovirus E1A enhancer-binding protein; AW414408; E1A F; E1AF; E1A-F; ETS translocation variant 4; ets variant 4; ets variant gene 4 (E1A enhancer binding protein, E1AF); ets variant gene 4 (E1A enhancer-binding protein, E1AF); ETS variant protein 4; Etv4; EWS protein/E1A enhancer binding protein chimera; PEA3; Pea-3; PEA3 protein; PEAS3; Polyomavirus enhancer activator 3; POLYOMAVIRUS ENHANCER ACTIVATOR 3 (PEA3 PROTEIN) (ETS TRANSLOCATION VARIANT 4); polyomavirus enhancer activator 3 homolog; polyomavirus enhancer activator-3; Protein PEA3 | |
| Rabbit | |
| Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
| RUO | |
| 18612, 2118 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P28322, P43268 | |
| ETV4 | |
| KLH conjugated synthetic peptide between 419-447 amino acids from the C-terminal region of human ETV4. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction