missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ETV5 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16696624
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16696624 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16696624 Supplier Invitrogen™ Supplier No. PA5100144

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ERM / Etv5.

ETS variant 5, transcription factor of the ETS family, divergent member of the winged helix-turn-helix superfamily, expressed in brain, placenta and other tissues involved in development. Tissue specificity: Ubiquitous. ERM is expressed in Sertoli cells and spermatogonial stem cells (SSCs). Studies on knockout mice have shown that it is required for SSC self-renewal but not differentiation, thus affected mice exhibit progressive germ cell depletion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ETV5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ETV5
Gene Accession No. P41161, Q9CXC9
Gene Alias 1110005E01Rik; 8430401F14Rik; ERM; ETS translocation variant 5; ets variant 5; ets variant gene 5; ets variant gene 5 (ets-related molecule); ets-related molecule; ets-related protein ERM; ETV5
Gene Symbols ETV5
Host Species Rabbit
Immunogen A synthesized peptide derived from human ETV5(Accession P41161), corresponding to amino acid residues P289-C339.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104156, 2119, 303828
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.