missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ETV5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA580602
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human ETV5.
ETS variant 5, transcription factor of the ETS family, divergent member of the winged helix-turn-helix superfamily, expressed in brain, placenta and other tissues involved in development. Tissue specificity: Ubiquitous. ERM is expressed in Sertoli cells and spermatogonial stem cells (SSCs). Studies on knockout mice have shown that it is required for SSC self-renewal but not differentiation, thus affected mice exhibit progressive germ cell depletion.
Specifications
| ETV5 | |
| Polyclonal | |
| Unconjugated | |
| ETV5 | |
| 1110005E01Rik; 8430401F14Rik; ERM; ETS translocation variant 5; ets variant 5; ets variant gene 5; ets variant gene 5 (ets-related molecule); ets-related molecule; ets-related protein ERM; ETV5 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 2119 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with no preservative | |
| P41161 | |
| ETV5 | |
| A synthetic peptide corresponding to the center region of the Human ETV5. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction