missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Mouse CXCL5/ENA-78 ELISA Kit (Colorimetric)

Product Code. 18238636 Shop All Bio Techne Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18238636

Brand: Novus Biologicals™ NBP278750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Assay Range 15.63 to 1000 pg/mL
Assay Sensitivity 9.38 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Other Biological Fluids, Plasma, Serum
Specificity This kit recognizes Mouse CXCL5 in samples. No Significant cross-reactivity or interference between Mouse CXCL5 and analogues was observed
For Use With (Application) ELISA
Gene ID (Entrez) 6374
Gene Symbol CXCL5
Host Species Mouse
Quantity 1 Kit
Research Discipline Biologically Active Proteins, Cell Cycle and Replication, Chemokines and Cytokines
Storage Requirements Storage of components varies. See protocol for specific instructions.
Target CXCL5/ENA-78
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.