missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETV7 Antibody (OTI3B2), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71938AF532
This item is not returnable.
View return policy
Description
ETV7 Monoclonal specifically detects ETV7 in Human samples. It is validated for Western Blot.
Specifications
| ETV7 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| ETV7 | |
| Recombinant protein fragment corresponding to amino acids 90-341 of human ETV7 (NP_057219) produced in E.coli. | |
| Primary | |
| Human | |
| IgG2b |
| Western Blot | |
| OTI3B2 | |
| Western Blot | |
| ETS translocation variant 7, ets variant 7, ets variant gene 7 (TEL2 oncogene), TEL-2, TEL2 oncogene, TEL2ETS-related protein Tel2, TELBEts transcription factor TEL-2b, Tel-related Ets factor, transcription factor ets, transcription factor ETV7, Transcription factor Tel-2, TREF | |
| Mouse | |
| 0.1 mL | |
| 51513 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction