missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETV7 Monoclonal Antibody (OTI3B2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA802936
This item is not returnable.
View return policy
Description
The ETS (E26 transformation-specific) family of transcription factors, of which ETV7 is a member, is a large group of evolutionarily conserved transcriptional regulators that play an important role in a variety of cellular processes throughout development and differentiation, and are involved in oncogenesis as well.Specifications
| ETV7 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y603 | |
| ETV7 | |
| Human recombit protein fragment corresponding to amino acids 90-341 of human ETV7 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| OTI3B2 | |
| Unconjugated | |
| ETV7 | |
| TEL-2, TEL2, TELB | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 51513 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction