missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETV7 Monoclonal Antibody (OTI3B2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA802936S
This item is not returnable.
View return policy
Description
ETV7 Monoclonal antibody specifically detects ETV7 in Human samples. It is validated for Western BlotSpecifications
| ETV7 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q969H0 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 55294 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI3B2 | |
| Unconjugated | |
| FBXW7 | |
| Ets transcription factor TEL-2b; ETS translocation variant 7; ETS variant 7; ets variant gene 7 (TEL2 oncogene); ETS-related protein Tel2; ETV7; TEL2; TEL-2; TEL2 oncogene; TELB; tel-related Ets factor; Transcription factor ETV7; Transcription factor Tel-2; TREF | |
| Human recombinant protein fragment corresponding to amino acids 1-354 of human FBXW7 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction