Learn More
Novus Biologicals™ EV MiniSEC Columns
Product Code. 30243672 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 Columns
20 Columns
Unit Size:
Pack of 10
Pack of 20
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30243672 20 Columns Pack of 20
30243673 10 Columns Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30243672 Supplier Novus Biologicals™ Supplier No. NBP34104320Columns
size exclusion chromatography (SEC) columns designed to isolate extracellular vesicles (EVs) from small volume samples
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at 4°C.
Quantity 20 Columns