Learn More
Heidolph™ Evaporating Cylinder
Product Code. 10279411 Shop All Heidolph Instruments Products
Change view
Click to view available options
Volume (Metric):
500 mL
1500 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10279411 500 mL Each
10468781 1500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10279411 Supplier Heidolph™ Supplier No. 5140007000
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
For use with rotary evaporators
Evaporating cylinders come with a standard NS 29/32 joint size but are also available in NS 24/29
For 500 ml filling capacity.
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Material Glass
Type Evaporating Cylinder
For Use With (Equipment) Rotary evaporators
Shape Cylinder
Volume (Metric) 500 mL