missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EVC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552591
This item is not returnable.
View return policy
Description
Immunogen sequence: GVMDLLEAQL ETQLQEAEQN FISELAALAR VPLAESKLLP AKRGLLEKPL RTKRKKPLPQ ERGDLGVPNN EDLASGDQTS GSLSSKRLSQ QESEAGDSGN SKKMLK Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 55%.
This gene encodes a protein containing a leucine zipper and a transmembrane domain. This gene has been implicated in both Ellis-van Creveld syndrome (EvC) and Weyers acrodental dysostosis.
Specifications
| EVC | |
| Polyclonal | |
| Unconjugated | |
| Evc | |
| DWF-1; Ellis van Creveld gene homolog; Ellis van Creveld gene syndrome; Ellis van Creveld protein; Ellis van Creveld syndrome; ellis-van Creveld syndrome protein; Ellis-van Creveld syndrome protein homolog; EVC; EvC ciliary complex subunit 1; EvC protein (Ellis-van Creveld syndrome); EVC1; EVCL | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2121 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P57679 | |
| Evc | |
| Recombinant protein corresponding to Human EVC. Recombinant protein control fragment (Product #RP-89974). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction