Learn More
EVI2B Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99056PCP
This item is not returnable.
View return policy
Description
EVI2B Polyclonal antibody specifically detects EVI2B in Mouse samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| EVI2B | |
| Western Blot, Immunoprecipitation | |
| PerCP | |
| Store at 4C in the dark. |