missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EWSR1 Antibody (5H7), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-92686AF405
This item is not returnable.
View return policy
Description
EWSR1 Monoclonal specifically detects EWSR1 in Human, Mouse, Rat, Canine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| EWSR1 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Q01844 | |
| EWSR1 | |
| Full length recombinant human EWSR1 expressed in and purified from E. coli. [UniProt# Q01844] | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine, Equine | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 5H7 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| bK984G1.4, Ewing sarcoma breakpoint region 1, Ewing sarcoma breakpoint region 1 protein, Ewings sarcoma EWS-Fli1 (type 1) oncogene, EWS oncogene, EWSRNA-binding protein EWS | |
| Mouse | |
| 85 kDa | |
| 0.1 mL | |
| Cancer, Cellular Markers, Neuroscience, Tumor Suppressors | |
| 2130 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction