missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EWSR1 Antibody (5H7), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-92686AF750
This item is not returnable.
View return policy
Description
EWSR1 Monoclonal specifically detects EWSR1 in Human, Mouse, Rat, Canine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| EWSR1 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| EWSR1 | |
| Full length recombinant human EWSR1 expressed in and purified from E. coli. [UniProtNo. Q01844] | |
| Primary | |
| Human, Mouse, Rat, Canine, Equine | |
| IgG2b |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 5H7 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| bK984G1.4, Ewing sarcoma breakpoint region 1, Ewing sarcoma breakpoint region 1 protein, Ewings sarcoma EWS-Fli1 (type 1) oncogene, EWS oncogene, EWSRNA-binding protein EWS | |
| Mouse | |
| 0.1 mL | |
| 2130 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction